missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Novus Biologicals™ Human Lung Whole Tissue Lysate (Adult Whole Pulmonary embolism)
Shop All Bio Techne ProductsDescription
The Human Lung Whole Tissue Lysate (Adult Whole Pulmonary embolism) is derived from human.
Specifications
Specifications
| Tissue | Respiratory |
| Quantity | 1 mg |
| Storage Buffer | HEPES (pH 7.9), MgCl2, KCl, EDTA, Sucrose, Glycerol, Sodium deoxycholate, NP-40, and a cocktail of protease inhibitors |
| Test Specificity | Total Protein - Pulmonary embolism: Lung |
| Cellular Fraction | Whole |
| Tissue Condition | Pulmonary embolism |
| Cross Reactivity | Human |
| Host Species | Human |
| Product Type | Human Lung Whole Tissue Lysate (Adult Pulmonary embolism) |
| Content And Storage | -80°C. Avoid freeze-thaw cycles. |
| Show More |
For Research Use Only
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?