missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Novus Biologicals™ Human Lung Whole Tissue Lysate (Adult Whole Normal)
Specifications
Specifications
| Preparation Method | Western Blot, Immunoprecipitation, Electrophoresis, Enzymatic activity analysis, Protein-protein interaction, Tissue specific expression |
| Quantity | 1 mg |
| Content And Storage | Store at -80°C. Avoid freeze/thaw cycles. |
| Target | Lung |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?