missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems Human Luminex™ Discovery Assay

Product Code. 18401330
Change view
Click to view available options
Quantity:
1 Plex
10 Plex
11 Plex
12 Plex
13 Plex
14 Plex
15 Plex
16 Plex
17 Plex
18 Plex
19 Plex
2 Plex
20 Plex
21 Plex
22 Plex
23 Plex
24 Plex
25 Plex
26 Plex
27 Plex
28 Plex
29 Plex
3 Plex
30 Plex
31 Plex
32 Plex
33 Plex
34 Plex
35 Plex
36 Plex
37 Plex
38 Plex
39 Plex
4 Plex
40 Plex
41 Plex
42 Plex
43 Plex
44 Plex
45 Plex
46 Plex
47 Plex
48 Plex
49 Plex
5 Plex
50 Plex
6 Plex
7 Plex
8 Plex
9 Plex
Unit Size:
1 set
Each
50 product options available for selection
Product selection table with 50 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18401330 39 Plex 1 set
30001637 17 Plex Each
30001583 14 Plex Each
30001543 2 Plex Each
18743167 9 Plex Each
18701207 5 Plex Each
30000711 4 Plex Each
18792114 19 Plex 1 set
18722114 13 Plex 1 set
18751994 18 Plex 1 set
18711994 10 Plex 1 set
18722464 12 Plex 1 set
18772204 8 Plex 1 set
18792234 15 Plex 1 set
18782234 11 Plex 1 set
18702204 22 Plex 1 set
18711924 7 Plex 1 set
18752044 16 Plex 1 set
18782144 20 Plex 1 set
18771844 1 Plex 1 set
18752434 21 Plex 1 set
18752284 6 Plex 1 set
18722004 3 Plex 1 set
18791834 35 Plex 1 set
18732014 32 Plex 1 set
18771944 23 Plex 1 set
18792174 33 Plex 1 set
18702174 26 Plex 1 set
18702104 42 Plex 1 set
18772054 28 Plex 1 set
18762294 27 Plex 1 set
18782364 31 Plex 1 set
18782214 30 Plex 1 set
18792094 25 Plex 1 set
18731834 43 Plex 1 set
18772324 24 Plex 1 set
18461770 45 Plex 1 set
18472310 40 Plex 1 set
18421230 37 Plex 1 set
18461520 49 Plex 1 set
18492540 44 Plex 1 set
18471520 34 Plex 1 set
18462550 50 Plex 1 set
18482310 47 Plex 1 set
18411530 48 Plex 1 set
18401230 29 Plex 1 set
18471730 38 Plex 1 set
18421790 41 Plex 1 set
18451260 46 Plex 1 set
18451350 36 Plex 1 set
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 50 options available.
50 options
This item is not returnable. View return policy
Product Code. 18401330 Supplier R&D Systems Supplier No. LXSAHM39

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Luminex bead-based multiplex assays are designed to provide accurate, reproducible results for every target analyte.

TRUSTED_SUSTAINABILITY

Specifications

Quantity 39 Plex
Species Human
Binding Target Multiplex Cytokine Assay Kits
Gene Symbol GZMB
Content And Storage Store the unopened product at 2 - 8 °C. Do not use past expiration date.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.