missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Human LUC7L (aa 312-339) Control Fragment Recombinant Protein
missing translation for 'orderingAttributeHoverText'
Quantity:
100 μL
missing translation for 'unitSize'
100µL
Beschreibung
Highest antigen sequence indentity to the following orthologs: Mouse (86%), Rat (86%). This recombinant protein control fragment may be used for blocking experiments with the corresponding antibody, PA5-66014 (PA5-66014. In IHC/ICC and WB experiments, we recommend a 100x molar excess of the protein fragment control based on the concentration and the molecular weight. Pre-incubate the antibody-protein control fragment mixture for 30 min at room temperature.
May bind to RNA via its Arg/Ser-rich domain.
Spezifikation
Spezifikation
| Accession Number | Q9NQ29 |
| Concentration | ≥5.0 mg/mL |
| For Use With (Application) | Blocking Assay, Control |
| Formulation | 1 M urea, PBS with no preservative; pH 7.4 |
| Gene ID (Entrez) | 55692 |
| Name | Human LUC7L (aa 312-339) Control Fragment |
| Quantity | 100 μL |
| Regulatory Status | RUO |
| Gene Alias | 1810045C04Rik; 2410018D03Rik; hLuc7B1; Luc7; Luc7 homolog (S. cerevisiae)-like; LUC7 like; LUC7B1; Luc7l; LUC7L1; LUC7-like; LUC7-like (S. cerevisiae); Putative RNA-binding protein Luc7-like 1; putative SR protein LUC7B1; sarcoplasmic reticulum protein LUC7B1; SR+89 |
| Common Name | LUC7L |
| Mehr anzeigen |
Berichtigung von Produktinhalten
Bitte geben Sie uns Ihr Feedback zu den Produktinhalten, indem Sie das folgende Formular ausfüllen.
Name des Produkts
Haben Sie Verbesserungsvorschläge?Übermitteln Sie eine inhaltliche Korrektur