missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human LTK Antibody, R&D Systems™
Sheep Polyclonal Antibody
Brand: R&D Systems AF4664-SP
This item is not returnable.
View return policy
Description
LTK Polyclonal specifically detects LTK in Human samples. It is validated for Western Blot, Immunohistochemistry.
Specifications
| LTK | |
| Polyclonal | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| EC 2.7.10.1, leukocyte receptor tyrosine kinase, leukocyte tyrosine kinase, leukocyte tyrosine kinase receptor, TYK1, TYK1Protein tyrosine kinase 1 | |
| Sheep | |
| Affinity Purified | |
| RUO | |
| 4058 | |
| Reconstitute at 0.2 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
| Western Blot, Immunohistochemistry | |
| LYOPH | |
| Western Blot 1 ug/mL, Immunohistochemistry 5-15 ug/mL | |
| P29376 | |
| LTK | |
| Mouse myeloma cell line NS0-derived recombinant human LTK Ser10-Pro424 Accession # P29376 | |
| 25 μg | |
| Primary | |
| Detects human LTK in direct ELISAs and Western blots. | |
| Human | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction