missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Human LTBP2 (aa 241-376) Control Fragment Recombinant Protein
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
Description
Highest antigen sequence indentity to the following orthologs: Mouse (60%), Rat (60%). This recombinant protein control fragment may be used for blocking experiments with the corresponding antibody, PA5-51930 (PA5-51930. In IHC/ICC and WB experiments, we recommend a 100x molar excess of the protein fragment control based on the concentration and the molecular weight. Pre-incubate the antibody-protein control fragment mixture for 30 min at room temperature.
The protein encoded by this gene belongs to the family of latent transforming growth factor (TGF)-beta binding proteins (LTBP), which are extracellular matrix proteins with multi-domain structure. This protein is the largest member of the LTBP family possessing unique regions and with most similarity to the fibrillins. It has thus been suggested that it may have multiple functions: as a member of the TGF-beta latent complex, as a structural component of microfibrils, and a role in cell adhesion.
Specifications
Specifications
| Accession Number | Q14767 |
| Concentration | ≥5.0 mg/mL |
| For Use With (Application) | Blocking Assay, Control |
| Formulation | 1 M urea, PBS with no preservative; pH 7.4 |
| Gene ID (Entrez) | 4053 |
| Name | Human LTBP2 (aa 241-376) Control Fragment |
| Quantity | 100 μL |
| Regulatory Status | RUO |
| Gene Alias | AW208642; C14orf141; GLC3D; latent transforming growth factor beta binding protein 2; latent-transforming growth factor beta-binding protein 2; LTBP2; LTBP-2; LTBP3; MSPKA; MSTP031; WMS3 |
| Common Name | LTBP2 |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction