missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Human LSM14A (aa 108-219) Control Fragment Recombinant Protein

Product Code. 30200335
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
This item is not returnable. View return policy

Product Code. 30200335

Brand: Invitrogen™ RP90714

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Recombinant Protein

Highest antigen sequence indentity to the following orthologs: Mouse (89%), Rat (89%). This recombinant protein control fragment may be used for blocking experiments with the corresponding antibody, PA5-53680 (PA5-53680. In IHC/ICC and WB experiments, we recommend a 100x molar excess of the protein fragment control based on the concentration and the molecular weight. Pre-incubate the antibody-protein control fragment mixture for 30 min at room temperature.

LSM14A, Sm-like proteins were identified in a variety of organisms based on sequence homology with the Sm protein family. Sm-like proteins contain the Sm sequence motif, which consists of 2 regions separated by a linker of variable length that folds as a loop. The Sm-like proteins are thought to form a stable heteromer present in tri-snRNP particles, which are important for pre-mRNA splicing.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number Q8ND56
Concentration ≥5.0 mg/mL
For Use With (Application) Blocking Assay, Control
Formulation 1 M urea, PBS with no preservative; pH 7.4
Gene ID (Entrez) 26065
Name Human LSM14A (aa 108-219) Control Fragment
Quantity 100 μL
Regulatory Status RUO
Gene Alias 2700023B17Rik; AA407828; AlphaSNBP; AU017544; C19orf13; FAM61A; family with sequence similarity 61, member A; hRAP55; hRAP55A; LSM14 homolog A; LSM14 homolog A (SCD6, S. cerevisiae); LSM14A; LSM14A mRNA processing body assembly factor; LSM14A, mRNA processing body assembly factor; LSM14A, SCD6 homolog A; LSM14A, SCD6 homolog A (S. cerevisiae); mRAP55A; Protein FAM61A; Protein LSM14 homolog A; protein SCD6 homolog; Putative alpha-synuclein-binding protein; RAP55; RAP55A; RGD1305695; RNA-associated protein 55; RNA-associated protein 55 A; Tral
Common Name LSM14A
Gene Symbol Lsm14a
Conjugate Unconjugated
Species Human
Recombinant Recombinant
Protein Tag His-ABP-tag
Sequence MGSYGPFGRMPTYSQFSPSSLVGQQFGAVGVAGSSLTSFGTETSNSGTLPQSSAVGSAFTQDTRSLKTQLSQGRSSPQLDPLRKSPTMEQAVQTASAHLPAPAAVGRRSPVS
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Expression System E. coli
Form Liquid
Purity or Quality Grade >80% by SDS-PAGE and Coomassie blue staining
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.