missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human LSECtin/CLEC4G Antibody, R&D Systems™

Goat Polyclonal Antibody
Brand: R&D Systems AF2947-SP
This item is not returnable.
View return policy
Description
LSECtin/CLEC4G Polyclonal specifically detects LSECtin/CLEC4G in Human samples. It is validated for Western Blot.
Specifications
| LSECtin/CLEC4G | |
| Polyclonal | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| CLEC4G, C-type lectin domain family 4 member G, C-type lectin domain family 4, member G, C-type lectin superfamily 4, member G, DTTR431, liver and lymph node sinusoidal endothelial cell C-type lectin, LP2698, LSECtin, Q6UXB4, UNQ431 | |
| Goat | |
| Affinity Purified | |
| RUO | |
| 339390 | |
| Reconstitute at 0.2 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
| Western Blot | |
| LYOPH | |
| Western Blot 0.1 ug/mL | |
| Q6UXB4 | |
| CLEC4G | |
| Mouse myeloma cell line NS0-derived recombinant human LSECtin/CLEC4G Ser54-Cys293 Accession # Q6UXB4 | |
| 25 μg | |
| Primary | |
| Detects human LSECtin/CLEC4G in direct ELISAs and Western blots. | |
| Human | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction