missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Human LSAMP (aa 216-262) Control Fragment Recombinant Protein

Product Code. 30205669
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
This item is not returnable. View return policy

Product Code. 30205669

Brand: Invitrogen™ RP106503

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Recombinant Protein

Highest antigen sequence indentity to the following orthologs: Mouse (100%), Rat (100%). This recombinant protein control fragment may be used for blocking experiments. In IHC/ICC and WB experiments, we recommend a 100x molar excess of the protein fragment control based on the concentration and the molecular weight. Pre-incubate the antibody-protein control fragment mixture for 30 min at room temperature.

The protein encoded by this gene is a neuronal surface glycoprotein found in cortical and subcortical regions of the limbic system. During development of the limbic system, this encoded protein is found on the surface of axonal membranes and growth cones, where it acts as a selective homophilic adhesion molecule, and guides the development of specific patterns of neuronal connections.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number Q13449
Concentration ≥5.0 mg/mL
For Use With (Application) Blocking Assay, Control
Formulation 1 M urea, PBS with no preservative; pH 7.4
Gene ID (Entrez) 4045
Name Human LSAMP (aa 216-262) Control Fragment
Quantity 100 μL
Regulatory Status RUO
Gene Alias 5430428I19; AW046674; B130007O04Rik; D930023J12Rik; IgLON family member 3; IGLON3; Lam; LAMP; limbic system associated membrane protein; limbic system-associated membrane protein; Lsamp; neuronal growth regulator 1-like; Unknown (protein for MGC:128495)
Common Name LSAMP
Gene Symbol Lsamp
Conjugate Unconjugated
Species Human
Recombinant Recombinant
Protein Tag His-ABP-tag
Sequence NYPPTITESKSNEATTGRQASLKCEASAVPAPDFEWYRDDTRINSAN
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Expression System E. coli
Form Liquid
Purity or Quality Grade >80% by SDS-PAGE and Coomassie blue staining
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.