missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human LRRK2 Antibody, R&D Systems™

Mouse Monoclonal Antibody
Brand: R&D Systems MAB6674-SP
This item is not returnable.
View return policy
Description
LRRK2 Monoclonal specifically detects LRRK2 in Human samples. It is validated for Immunohistochemistry.
Specifications
| LRRK2 | |
| Monoclonal | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. | |
| augmented in rheumatoid arthritis 17, AURA17, Dardarin, DKFZp434H2111, EC 2.7.11.1, FLJ45829, leucine-rich repeat kinase 2, leucine-rich repeat serine/threonine-protein kinase 2, PARK8, PARK8DARDARIN, Parkinson disease (autosomal dominant) 8, RIPK7, ROCO2 | |
| E. coli-derived recombinant human LRRK2, Val241-Val500, Accession No. Q5S007 | |
| Primary | |
| Detects human LRRK2 in direct ELISAs. | |
| Human | |
| IgG2a |
| Immunohistochemistry | |
| 521313 | |
| Immunohistochemistry 5-25 μ/mL | |
| Q5S007 | |
| Mouse | |
| 25 μg | |
| 120892 | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70°C as supplied. 1 month, 2 to 8°C under sterile conditions after reconstitution. 6 months, -20 to -70°C under sterile conditions after reconstitution. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction