missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Human LRRFIP2 Synthetic Peptide

Product Code. 15265229
Change view
Click to view available options
Quantity:
50 μg
Unit Size:
50µg
Product Code. Quantity unitSize
15265229 50 μg 50µg
1 options
This item is not returnable. View return policy

Product Code. 15265229

Brand: Invitrogen™ PEP1229

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Synthetic Peptide

LRRFIP2 Synthetic Peptide, PEP-1229, from Invitrogen.

The leucine-rich repeat FLI-I-interacting protein 2 (LRRFIP2), like the related protein LRRFIP1, was identified in a yeast two-hybrid system through binding to the LRR domain of human FLI. It can activate the Wnt signaling pathway in cultured cells and is thought to be a component of the Wnt signaling pathway that modulates Wnt signaling through interactions with Disheveled to increase the cellular levels and transcription activity of beta-catenin. LRRFIP2 has recently been characterized as a positive regulator of the TLR4 signaling pathway for activating NF-kappa-B during the early host response to LPS stimulation through binding to the TLR adaptor protein MyD88, and that this interaction with MyD88 is governed by phosphorylation of specific residues in LRRFIP2.
TRUSTED_SUSTAINABILITY

Specifications

Gene ID (Entrez) 9209
Species Human
Purification Method purified
Accession Number Q9Y608
Common Name Human LRRFIP2
Concentration 200 μg/mL
Content And Storage -20°C
Description Synthetic Peptide
Format Liquid
Formulation PBS with 0.1% BSA and 0.02% sodium azide; pH 7.2
For Use With (Application) Blocking Assay, Control
Gene Symbol LRRFIP2
Lyophilized No
Preservative 0.02% sodium azide
Product Type Peptide
Quantity 50 μg
Regulatory Status RUO - research use only
Storage Buffer PBS
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.