missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Human LRRFIP2 (aa 237-324) Control Fragment Recombinant Protein

Product Code. 30196268
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
This item is not returnable. View return policy

Product Code. 30196268

Brand: Invitrogen™ RP96320

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Recombinant Protein

Highest antigen sequence indentity to the following orthologs: Mouse (90%), Rat (90%). This recombinant protein control fragment may be used for blocking experiments with the corresponding antibody, PA5-57481 (PA5-57481. In IHC/ICC and WB experiments, we recommend a 100x molar excess of the protein fragment control based on the concentration and the molecular weight. Pre-incubate the antibody-protein control fragment mixture for 30 min at room temperature.

The leucine-rich repeat FLI-I-interacting protein 2 (LRRFIP2), like the related protein LRRFIP1, was identified in a yeast two-hybrid system through binding to the LRR domain of human FLI. It can activate the Wnt signaling pathway in cultured cells and is thought to be a component of the Wnt signaling pathway that modulates Wnt signaling through interactions with Disheveled to increase the cellular levels and transcription activity of beta-catenin. LRRFIP2 has recently been characterized as a positive regulator of the TLR4 signaling pathway for activating NF-kappa-B during the early host response to LPS stimulation through binding to the TLR adaptor protein MyD88, and that this interaction with MyD88 is governed by phosphorylation of specific residues in LRRFIP2.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number Q9Y608
Concentration ≥5.0 mg/mL
For Use With (Application) Blocking Assay, Control
Formulation 1 M urea, PBS with no preservative; pH 7.4
Gene ID (Entrez) 9209
Name Human LRRFIP2 (aa 237-324) Control Fragment
Quantity 100 μL
Regulatory Status RUO
Gene Alias 5133400F20Rik; AI850587; HUFI-2; leucine rich repeat (in FLII) interacting protein 2; leucine-rich repeat flightless-interactin protein 2-like protein; leucine-rich repeat flightless-interacting protein 2; LRR binding FLII interacting protein 2; LRR FLII-interacting protein 2; LRRFIP2
Common Name LRRFIP2
Gene Symbol LRRFIP2
Conjugate Unconjugated
Species Human
Recombinant Recombinant
Protein Tag His-ABP-tag
Sequence PGFTNDDTASIVSSDRASRGRRESVVSAADYFSRSNRRGSVVSEVDDISIPDLSSLDEKSDKQYAENYTRPSSRNSASATTPLSGNSS
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Expression System E. coli
Form Liquid
Purity or Quality Grade >80% by SDS-PAGE and Coomassie blue staining
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.