missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Human LRRC6 (aa 370-451) Control Fragment Recombinant Protein

Product Code. 30196837
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
This item is not returnable. View return policy

Product Code. 30196837

Brand: Invitrogen™ RP93781

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Recombinant Protein

Highest antigen sequence indentity to the following orthologs: Mouse (69%), Rat (69%). This recombinant protein control fragment may be used for blocking experiments with the corresponding antibody, PA5-55756 (PA5-55756. In IHC/ICC and WB experiments, we recommend a 100x molar excess of the protein fragment control based on the concentration and the molecular weight. Pre-incubate the antibody-protein control fragment mixture for 30 min at room temperature.

LRRC6 may be involved in spermatocytogenesis or prophase of meiosis.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number Q86X45
Concentration ≥5.0 mg/mL
For Use With (Application) Blocking Assay, Control
Formulation 1 M urea, PBS with no preservative; pH 7.4
Gene ID (Entrez) 23639
Name Human LRRC6 (aa 370-451) Control Fragment
Quantity 100 μL
Regulatory Status RUO
Gene Alias CILD19; leucine rich repeat containing 6; leucine rich repeat containing 6 (testis); leucine-rich repeat-containing 6 (testis); leucine-rich repeat-containing protein 6; leucine-rich repeat-containing protein 6; protein tilB homolog; leucine-rich testis-specific protein; LRRC6; Lrtp; Mc2; Protein tilB homolog; testis-specific leucine-rich repeat protein; Tslrp; Unknown (protein for MGC:138048)
Common Name LRRC6
Gene Symbol LRRC6
Conjugate Unconjugated
Species Human
Recombinant Recombinant
Protein Tag His-ABP-tag
Sequence TTGHLVICMPKVGEVITGGQRAFKSMKTTSDRSREQTNTRSKHMEKLEVDPSKHSFPDVTNIVQEKKHTPRRRPEPKIIPSE
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Expression System E. coli
Form Liquid
Purity or Quality Grade >80% by SDS-PAGE and Coomassie blue staining
Mostrar más Mostrar menos
Corrección del contenido de un producto

Proporcione sus comentarios sobre el contenido del producto rellenando el siguiente formulario.

Título del producto

Al hacer clic en Enviar, acepta que Fisher Scientific se ponga en contacto con usted en relación con los comentarios que ha proporcionado en este formulario. No compartiremos su información para ningún otro fin. Toda la información de contacto proporcionada se mantendrá de acuerdo con nuestra Política de Privacidad. Política de privacidad.

Gracias por ayudarnos a mejorar nuestra web. Su comentario ha sido enviado