missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Human LRRC4 ELISA Kit

Product Code. 17135079
Click to view available options
Quantity:
10 x 96 Tests
5 x 96 Tests
96 Tests
Unit Size:
Each
Pack of 10
Pack of 96
This item is not returnable. View return policy

Product Code. 17135079

Brand: Invitrogen™ EH307RBX5

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

ELISA

Human LRRC4 quantitates human LRRC4 in serum, plasma, supernatant. The assay will exclusively recognize both natural and recombinant human LRRC4.

LRRC4 protein function is currently unknown.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number BAG; brain tumor associated protein LRRC4; Brain tumor-associated protein BAG; brain tumor-associated protein MBAG1; leucine rich repeat containing 4; leucine rich repeat containing 4 protein; leucine-rich repeat-containing 4; leucine-rich repeat-containing protein 4; LIBG-like protein; LRRC4; LRRC4-like protein; MBAG1; NAG14; Nasopharyngeal carcinoma-associated gene 14 protein; Netrin-G2 ligand; NGL-2; NLG2; NLG-2; protein NGL-2; UNQ554/PRO1111
Assay Range 0.42 to 100 ng/mL
Assay Sensitivity 0.42 ng/mL
Conjugate HRP
Product Type ELISA
Sample Type Plasma, Serum, Supernatant
For Use With (Equipment) Colorimetric Microplate Reader
Gene ID (Entrez) 64101
Gene Symbol Lrrc4
Interassay CV <12%
Intraassay CV <10
Kit Contents Pre-coated 96 well plate, Standard, Assay Diluent concentrate, Biotinylated Detection Antibody, SAV-HRP, Wash Buffer, Chromogen, Stop Solution, Adhesive Plate Covers
Label or Dye HRP
Quantity 5 x 96 Tests
Regulatory Status RUO
Sample Volume Plasma 50 μL, Serum 50 μL, Supernatant 50 μL
Storage Requirements 2°C to 8°C
Target LRRC4
Test Time 1 hrs 20 mins
Total Assay Time 4 hrs 45 mins
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.