missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human LRPAP Antibody, R&D Systems™

Goat Polyclonal Antibody
Brand: R&D Systems AF4296-SP
This item is not returnable.
View return policy
Description
LRPAP Polyclonal specifically detects LRPAP in Human samples. It is validated for Western Blot, Immunocytochemistry.
Specifications
| LRPAP | |
| Polyclonal | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| A2MRAP, A2MRAPMRAP, A2RAP, alpha-2-macroglobulin receptor-associated protein, alpha-2-macroglobulin receptor-associated protein 1, Alpha-2-MRAP, HBP44, lipoprotein receptor associated protein, low density lipoprotein receptor-related protein associated protein 1, Low density lipoprotein receptor-related protein-associated protein 1, low density lipoprotein-related protein-associated protein 1(alpha-2-macroglobulin receptor-associated protein 1), LRPAP1, MGC138272, RAP | |
| Goat | |
| Affinity Purified | |
| RUO | |
| 4043 | |
| Reconstitute at 0.2 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
| Western Blot, Immunocytochemistry | |
| LYOPH | |
| Western Blot 0.1 ug/mL, Immunocytochemistry 5-15 ug/mL | |
| P30533 | |
| LRPAP1 | |
| E. coli-derived recombinant human LRPAP Tyr35-Leu357 Accession # P30533 | |
| 25 μg | |
| Primary | |
| Detects human LRPAP in direct ELISAs and Western blots. In Western blots, approximately 30% cross-reactivity with recombinant mouse LRPAP is observed. | |
| Human | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction