missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human LRP-6 Alexa Fluor™ 350-conjugated Antibody, R&D Systems™

Mouse Monoclonal Antibody
Brand: R&D Systems FAB1505U-100UG
This item is not returnable.
View return policy
Description
LRP-6 Monoclonal specifically detects LRP-6 in Human samples. It is validated for Flow Cytometry.
Specifications
| LRP-6 | |
| Monoclonal | |
| 0.2 mg/mL | |
| Flow Cytometry 0.25-1 ug/10^6 cells | |
| O75581 | |
| LRP6 | |
| Mouse myeloma cell line NS0-derived recombinant human LRP-6 Ala20-Pro1368 Accession # O75581 | |
| RUO | |
| 4040 | |
| Human | |
| IgG2a |
| Flow Cytometry | |
| 255302 | |
| Alexa Fluor 350 | |
| Supplied 0.2 mg/mL in a saline solution containing BSA and Sodium Azide. with 0.09% Sodium Azide | |
| ADCAD2, FLJ90062, FLJ90421, low density lipoprotein receptor-related protein 6, low-density lipoprotein receptor-related protein 6, LRP6, LRP-6 | |
| Mouse | |
| 100 μg | |
| Primary | |
| Detects human LRP-6 in direct ELISAs. In direct ELISAs, approximately 5% cross-reactivity with recombinant mouse (rm) LRP-6 and no cross-reactivity with recombinant human (rh) LRP-1, -4, -5, rhLRP-6 (intracellular domain), or rhLDL R is observed. | |
| Store the unopened product at 2 - 8 degreesC. Do not use past expiration date. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only (RUO)
Spot an opportunity for improvement?Share a Content Correction