missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human LRP-1 Cluster II Antibody, R&D Systems™

Goat Polyclonal Antibody has been used in 2 publications
Brand: R&D Systems AF2368-SP
This item is not returnable.
View return policy
Description
LRP-1 Cluster II Polyclonal specifically detects LRP-1 Cluster II in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry.
Specifications
| LRP-1 Cluster II | |
| Polyclonal | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| LRP1 | |
| Mouse myeloma cell line NS0-derived recombinant human LRP-1 Cluster II Arg786-Leu1165 Accession # Q07954 | |
| 25 μg | |
| Primary | |
| Detects human LRP-1 Cluster II in direct ELISAs and Western blots. In direct ELISAs, less than 2% cross-reactivity with recombinant human (rh) CRP-6 and rhMegalin-C4 is observed. | |
| Human | |
| IgG |
| Western Blot | |
| LYOPH | |
| Western Blot 0.1 ug/mL | |
| Q07954 | |
| Goat | |
| Affinity Purified | |
| RUO | |
| 4035 | |
| Reconstitute at 0.2 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction