missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human LRP-1 Cluster II Antibody, R&D Systems™
Goat Polyclonal Antibody has been used in 2 publications
Brand: R&D Systems AF2368
This item is not returnable.
View return policy
Description
LRP-1 Cluster II Polyclonal specifically detects LRP-1 Cluster II in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry.
Specifications
| LRP-1 Cluster II | |
| Polyclonal | |
| Unconjugated | |
| Q07954 | |
| Goat | |
| Affinity Purified | |
| RUO | |
| 4035 | |
| Reconstitute at 0.2 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
| Western Blot | |
| LYOPH | |
| Western Blot 0.1 ug/mL | |
| LRP1 | |
| Mouse myeloma cell line NS0-derived recombinant human LRP-1 Cluster II Arg786-Leu1165 Accession # Q07954 | |
| 100 μg | |
| Primary | |
| Detects human LRP-1 Cluster II in direct ELISAs and Western blots. In direct ELISAs, less than 2% cross-reactivity with recombinant human (rh) CRP-6 and rhMegalin-C4 is observed. | |
| Human | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction