missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Human LRFN3 (aa 374-415) Control Fragment Recombinant Protein

Product Code. 30199113
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
This item is not returnable. View return policy

Product Code. 30199113

Brand: Invitrogen™ RP100946

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Recombinant Protein

Highest antigen sequence indentity to the following orthologs: Mouse (98%), Rat (98%). This recombinant protein control fragment may be used for blocking experiments with the corresponding antibody, PA5-61839 (PA5-61839. In IHC/ICC and WB experiments, we recommend a 100x molar excess of the protein fragment control based on the concentration and the molecular weight. Pre-incubate the antibody-protein control fragment mixture for 30 min at room temperature.

LRFN3 is one of a family of five transmembrane glycoproteins that are highly expressed in neuronal tissues. LRFN proteins share leucine-rich repeat (LRR)-immunoglobulin-like (Ig)-fibronectin type III (Fn)-transmembrane domain structure with other members of the LRR-Ig-Fn protein superfamily such as the Slitrk family of proteins. Expression of LRFN1, -3, and -4 mRNA was detected in embryonic neuronal cells, while Lrfn2 and Lrfn5 expression was primarily restricted to more mature cells. LRFN1, -2, and -4 bound to PDZ domains of postsynaptic PSD95, re-distributing PSD95 to the cell periphery. It has been suggested that the Lrfn proteins play a role in the developing and/or mature vertebrate nervous system.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number Q9BTN0
Concentration ≥5.0 mg/mL
For Use With (Application) Blocking Assay, Control
Formulation 1 M urea, PBS with no preservative; pH 7.4
Gene ID (Entrez) 79414
Name Human LRFN3 (aa 374-415) Control Fragment
Quantity 100 μL
Regulatory Status RUO
Gene Alias A530045B06Rik; EMBL:AAI58863.1, ECO:0000312; fibronectin type III, immunoglobulin and leucine rich repeat domains 1; FIGLER1; leucine rich repeat and fibronectin type III domain containing 3; leucine-rich repeat and fibronectin type-III domain-containing protein 3; LRFN3; lrfn3 {ECO:0000312; PubMed:18227064}; RGD:1305567}; Salm4; Synaptic adhesion-like molecule 4; synaptic adhesion-like molecule 4 {ECO:0000303; UNQ5865/PRO34192
Common Name LRFN3
Gene Symbol LRFN3
Conjugate Unconjugated
Species Human
Recombinant Recombinant
Protein Tag His-ABP-tag
Sequence EATAAVELTVGPPPPPQLANSTSCDPPRDGDPDALTPPSAAS
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Expression System E. coli
Form Liquid
Purity or Quality Grade >80% by SDS-PAGE and Coomassie blue staining
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.