Learn More
Abnova™ Human LRCH4 Partial ORF (NP_002310.2, 274 a.a. - 379 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Brand: Abnova™ H00004034-Q01.25ug
Additional Details : Weight : 0.02000kg
Description
This gene encodes a protein that contains leucine-rich repeats (LRR) at its amino terminus and that is known to be involved in ligand binding. The carboxyl terminus may act as a membrane anchor. Identified structural elements suggest that the encoded protein resembles a receptor. [provided by RefSeq]
Sequence: PSRPPSFSPCPAEDLFPGHRYDGGLDSGFHSVDSGSKRWSGNESTDEFSELSFRISELAREPRGPRERKEDGSADGDPVQIDFIDSHVPGEDEERGTVEEQRPPELSpecifications
NP_002310.2 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
37.29kDa | |
12.5% SDS-PAGE Stained with Coomassie Blue | |
PSRPPSFSPCPAEDLFPGHRYDGGLDSGFHSVDSGSKRWSGNESTDEFSELSFRISELAREPRGPRERKEDGSADGDPVQIDFIDSHVPGEDEERGTVEEQRPPEL | |
RUO | |
LRCH4 | |
Wheat Germ (in vitro) | |
GST | |
Liquid |
Antibody Production, ELISA, Protein Array, Western Blot | |
4034 | |
LRCH4 (Human) Recombinant Protein (Q01) | |
25 ug | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
FLJ40101/FLJ46315/LRN/LRRN1/LRRN4/PP14183/SAP25 | |
LRCH4 | |
Recombinant | |
wheat germ expression system |