missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Human LPPR5 (aa 282-321) Control Fragment Recombinant Protein

Product Code. 30199557
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
This item is not returnable. View return policy

Product Code. 30199557

Brand: Invitrogen™ RP91257

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Recombinant Protein

Highest antigen sequence indentity to the following orthologs: Mouse (80%), Rat (80%). This recombinant protein control fragment may be used for blocking experiments with the corresponding antibody, PA5-53715 (PA5-53715. In IHC/ICC and WB experiments, we recommend a 100x molar excess of the protein fragment control based on the concentration and the molecular weight. Pre-incubate the antibody-protein control fragment mixture for 30 min at room temperature.

The protein encoded by this gene is a type 2 member of the phosphatidic acid phosphatase (PAP) family. All type 2 members of this protein family contain 6 transmembrane regions, and a consensus N-glycosylation site. PAPs convert phosphatidic acid to diacylglycerol, and function in de novo synthesis of glycerolipids as well as in receptor-activated signal transduction mediated by phospholipase D. Alternate transcriptional splice variants, encoding different isoforms, have been characterized. [provided by RefSeq, Jul 2008] Induces filopodia formation and promotes neurite growth in a CDC42-independent manner; impedes neurite growth inhibitory-mediated axonal retraction.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number Q32ZL2
Concentration ≥5.0 mg/mL
For Use With (Application) Blocking Assay, Control
Formulation 1 M urea, PBS with no preservative; pH 7.4
Gene ID (Entrez) 163404
Name Human LPPR5 (aa 282-321) Control Fragment
Quantity 100 μL
Regulatory Status RUO
Gene Alias 4833424O15Rik; Lipid phosphate phosphatase-related protein type 5; Lppr5; PAP2; Pap2d; phosphatidic acid phosphatase 2 d; phosphatidic acid phosphatase type 2; phosphatidic acid phosphatase type 2 d; phospholipid phosphatase related 5; phospholipid phosphatase-related protein type 5; Plasticity-related gene 5 protein; plasticity-related protein 5; PLPPR5; PRG5; PRG-5
Common Name LPPR5
Gene Symbol PLPPR5
Conjugate Unconjugated
Species Human
Recombinant Recombinant
Protein Tag His-ABP-tag
Sequence EHIHMDNLAQMPMISIPRVESPLEKVTSVQNHITAFAEVT
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Expression System E. coli
Form Liquid
Purity or Quality Grade >80% by SDS-PAGE and Coomassie blue staining
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.