missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human LPAR1/LPA1/EDG-2 Antibody, R&D Systems™
Description
LPAR1/LPA1/EDG-2 Monoclonal specifically detects LPAR1/LPA1/EDG-2 in Human samples. It is validated for Flow Cytometry, CyTOF-ready.
Specifications
Specifications
| Antigen | LPAR1/LPA1/EDG-2 |
| Applications | Flow Cytometry, CyTOF |
| Classification | Monoclonal |
| Clone | 1020714 |
| Conjugate | Unconjugated |
| Dilution | Flow Cytometry 0.25 ug/10^6 cells, CyTOF-ready |
| Formulation | Lyophilized from a 0.2 µm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 µm filtered solution in PBS. |
| Gene Accession No. | Q92633 |
| Gene Alias | edg-2, EDG2, EDG2Lysophosphatidic acid receptor Edg-2, endothelial differentiation, lysophosphatidic acid G-protein-coupled receptor2,, GPCR26, GPR26, LPA receptor 1, LPA1, LPA-1, LPA1vzg-1, lysophosphatidic acid receptor 1, Mrec1.3, rec.1.3, ventricular zone gene 1, vzg-1, VZG1 |
| Host Species | Mouse |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?