missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human LOX-1/OLR1 Antibody, R&D Systems™

Mouse Monoclonal Antibody
Brand: R&D Systems MAB17981-100
This item is not returnable.
View return policy
Description
LOX-1/OLR1 Monoclonal specifically detects LOX-1/OLR1 in Human samples. It is validated for Blockade of Receptor-ligand Interaction, ELISA Capture (Matched Antibody Pair).
Specifications
| LOX-1/OLR1 | |
| Monoclonal | |
| LYOPH | |
| Blockade of Receptor-ligand Interaction, ELISA Capture (Matched Antibody Pair) 2-8 ug/mL | |
| P78380 | |
| OLR1 | |
| Mouse myeloma cell line NS0-derived recombinant human LOX-1 Ser61-Gln273 Accession # P78380 | |
| 100 μg | |
| Primary | |
| Detects human LOX-1 in ELISAs. In ELISAs, this antibody does not cross-react with recombinant mouse LOX-1. | |
| Human | |
| Supernatant |
| Blockade of Receptor-ligand Interaction, ELISA | |
| 331219 | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| CLEC8A, CLEC8ASLOX1, C-type lectin domain family 8 member A, hLOX-1, Lectin-like oxidized LDL receptor 1, Lectin-like oxLDL receptor 1, Lectin-type oxidized LDL receptor 1, LOX1, LOX-1, LOX1ox LDL receptor 1, LOXIN, OLR1, oxidised low density lipoprotein (lectin-like) receptor 1, oxidized low density lipoprotein (lectin-like) receptor 1, oxidized low-density lipoprotein receptor 1, oxidized low-density lipoprotein receptor 1, soluble form, Ox-LDL receptor 1, SCARE1, scavenger receptor class E, member 1, SR-E1 | |
| Mouse | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| 4973 | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction