missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Human LOH12CR1 (aa 123-196) Control Fragment Recombinant Protein

Product Code. 30182760
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
This item is not returnable. View return policy

Product Code. 30182760

Brand: Invitrogen™ RP97841

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Recombinant Protein

Highest antigen sequence indentity to the following orthologs: Mouse (89%), Rat (89%). This recombinant protein control fragment may be used for blocking experiments with the corresponding antibody, PA5-58821 (PA5-58821. In IHC/ICC and WB experiments, we recommend a 100x molar excess of the protein fragment control based on the concentration and the molecular weight. Pre-incubate the antibody-protein control fragment mixture for 30 min at room temperature.

LOH12CR1 is a protein coding gene.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number Q969J3
Concentration ≥5.0 mg/mL
For Use With (Application) Blocking Assay, Control
Formulation 1 M urea, PBS with no preservative; pH 7.4
Gene ID (Entrez) 118426
Name Human LOH12CR1 (aa 123-196) Control Fragment
Quantity 100 μL
Regulatory Status RUO
Gene Alias 5830457J20Rik; AW124643; BLOC-1 related complex subunit 5; BLOC-1-related complex subunit 5; BORCS5; Loh12cr1; LOH1CR12; Loss of heterozygosity 12 chromosomal region 1; loss of heterozygosity 12 chromosomal region 1 protein; loss of heterozygosity 12 chromosomal region 1 protein homolog; loss of heterozygosity, 12, chromosomal region 1; loss of heterozygosity, 12, chromosomal region 1 homolog (human); Myristoylated lysosomal protein; Myrlysin
Common Name LOH12CR1
Gene Symbol BORCS5
Conjugate Unconjugated
Species Human
Recombinant Recombinant
Protein Tag His-ABP-tag
Sequence LSVETLFSFMQERQKRYAKYAEQIQKVNEMSAILRRIQMGIDQTVPLLDRLNSMLPEGERLEPFSMKPDRELRL
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Expression System E. coli
Form Liquid
Purity or Quality Grade >80% by SDS-PAGE and Coomassie blue staining
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.