missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Human LNPEP Synthetic Peptide
Click to view available options
Quantity:
50 μg
Unit Size:
50µg
Description
LNPEP Synthetic Peptide, PEP-1500, from Invitrogen.
This gene encodes a zinc-dependent aminopeptidase that cleaves vasopressin, oxytocin, lys-bradykinin, met-enkephalin, dynorphin A and other peptide hormones. The protein can be secreted in maternal serum, reside in intracellular vesicles with the insulin-responsive glucose transporter GLUT4, or form a type II integral membrane glycoprotein. The protein catalyzes the final step in the conversion of angiotensinogen to angiotensin IV (AT4) and is also a receptor for AT4. Alternative splicing results in multiple transcript variants encoding different isoforms.
Specifications
Specifications
| Gene ID (Entrez) | 4012 |
| Species | Human |
| Purification Method | purified |
| Accession Number | Q9UIQ6 |
| Common Name | Human LNPEP |
| Concentration | 200 μg/mL |
| Content And Storage | -20°C |
| Description | Synthetic Peptide |
| Format | Liquid |
| Formulation | PBS with 0.1% BSA and 0.02% sodium azide; pH 7.2 |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction