missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human LMTK2 Antibody, R&D Systems™

Rat Monoclonal Antibody
Brand: R&D Systems MAB5147-SP
This item is not returnable.
View return policy
Description
LMTK2 Monoclonal specifically detects LMTK2 in Human samples. It is validated for Western Blot.
Specifications
| LMTK2 | |
| Monoclonal | |
| LYOPH | |
| Western Blot 1 ug/mL | |
| Q8IWU2 | |
| LMTK2 | |
| E. coli-derived recombinant human LMTK2 Ala93-Tyr284 Accession # Q8IWU2 | |
| 25 μg | |
| Primary | |
| Detects human LMTK2 in direct ELISAs and Western blots. In direct ELISAs and Western blots, no cross-reactivity with recombinant human AATK is observed. | |
| Human | |
| Purified |
| Western Blot | |
| 465222 | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| AATK2, AATYK2FLJ46659, Apoptosis-associated tyrosine kinase 2, Brain-enriched kinase, BREK, BREKCDK5/p35-regulated kinase, cprk, EC 2.7.11, KIAA1079hBREK, kinase phosphatase inhibitor 2, Kinase/phosphatase/inhibitor 2, KPI-2, KPI2EC 2.7.11.1, lemur tyrosine kinase 2Serine/threonine-protein kinase KPI-2, LMR2, LMR2cyclin-dependent kinase 5/p35-regulated kinase, serine/threonine-protein kinase LMTK2 | |
| Rat | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| 22853 | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
| IgG2b |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction