missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human LMP7/PSMB8 Antibody, R&D Systems™

Mouse Monoclonal Antibody
Brand: R&D Systems MAB7710-SP
This item is not returnable.
View return policy
Description
LMP7/PSMB8 Monoclonal specifically detects LMP7/PSMB8 in Human samples. It is validated for Western Blot.
Specifications
| LMP7/PSMB8 | |
| Monoclonal | |
| LYOPH | |
| Western Blot 2 ug/mL | |
| P28062 | |
| PSMB8 | |
| E. coli-derived recombinant human LMP7/PSMB8 Thr73-Gln276 Accession # P28062 | |
| 25 μg | |
| Primary | |
| Detects human LMP7/PSMB8 in ELISAs. In direct ELISAs, no cross-reactivity with recombinant human LMPX/PSMB5 is observed. | |
| Human | |
| Purified |
| Western Blot | |
| 832913 | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| ALDD, beta5i, D6S216E, EC 3.4.25.1, JMP, LMP7MGC1491, Low molecular mass protein 7, low molecular weight protein 7, Macropain subunit C13, Multicatalytic endopeptidase complex subunit C13, NKJO, protease component C13, proteasome (prosome, macropain) subunit, beta type, 8 (large multifunctionalpeptidase 7), proteasome catalytic subunit 3i, Proteasome component C13, proteasome subunit beta 5i, proteasome subunit beta type-8, Proteasome subunit beta-5i, proteasome subunit Y2, proteasome-related gene 7, PSMB5i, PSMB5iD6S216, PSMB8, Really interesting new gene 10 protein, RING10, RING10proteasome (prosome, macropain) subunit, beta type, 8 (large multifunctionalprotease 7), Y2 | |
| Mouse | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| 5696 | |
| Sterile PBS to a final concentration of 0.5 mg/mL. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
| IgG2b |
Corrección del contenido de un producto
Proporcione sus comentarios sobre el contenido del producto rellenando el siguiente formulario.
Título del producto
For Research Use Only
¿Detecta una oportunidad de mejora?Comparta una corrección de contenido