missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Human LMAN2L (aa 170-307) Control Fragment Recombinant Protein
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
Description
Highest antigen sequence indentity to the following orthologs: Mouse (93%), Rat (93%). This recombinant protein control fragment may be used for blocking experiments with the corresponding antibody, PA5-55355 (PA5-55355. In IHC/ICC and WB experiments, we recommend a 100x molar excess of the protein fragment control based on the concentration and the molecular weight. Pre-incubate the antibody-protein control fragment mixture for 30 min at room temperature.
May be involved in the regulation of export from the endoplasmic reticulum of a subset of glycoproteins. May function as a regulator of ERGIC-53.
Specifications
Specifications
| Accession Number | Q9H0V9 |
| Concentration | ≥5.0 mg/mL |
| For Use With (Application) | Blocking Assay, Control |
| Formulation | 1 M urea, PBS with no preservative; pH 7.4 |
| Gene ID (Entrez) | 81562 |
| Name | Human LMAN2L (aa 170-307) Control Fragment |
| Quantity | 100 μL |
| Regulatory Status | RUO |
| Gene Alias | A630028F14Rik; Lectin mannose-binding 2-like; lectin, mannose binding 2 like; lectin, mannose-binding 2-like; LMAN2L; LMAN2-like protein; MRT52; PSEC0028; UNQ368/PRO704; VIP36-like; VIP36-like protein; VIPL |
| Common Name | LMAN2L |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction