missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Human LIPM (aa 33-68) Control Fragment Recombinant Protein
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
Description
Highest antigen sequence indentity to the following orthologs: Mouse (81%), Rat (81%). This recombinant protein control fragment may be used for blocking experiments with the corresponding antibody, PA5-145001 (PA5-145001. In IHC/ICC and WB experiments, we recommend a 100x molar excess of the protein fragment control based on the concentration and the molecular weight. Pre-incubate the antibody-protein control fragment mixture for 30 min at room temperature.
Plays a highly specific role in the last step of keratinocyte differentiation. May have an essential function in lipid metabolism of the most differentiated epidermal layers.
Specifications
Specifications
| Accession Number | Q5VYY2 |
| Concentration | ≥5.0 mg/mL |
| For Use With (Application) | Blocking Assay, Control |
| Formulation | 1 M urea, PBS with no preservative; pH 7.4 |
| Gene ID (Entrez) | 340654 |
| Name | Human LIPM (aa 33-68) Control Fragment |
| Quantity | 100 μL |
| Regulatory Status | RUO |
| Gene Alias | bA304I5.1; lipase family member M; lipase M; lipase member M; lipase, family member M; lipase-like abhydrolase domain-containing protein 3; lipase-like, ab-hydrolase domain containing 3; LIPL3; LIPM |
| Common Name | LIPM |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction