missing translation for 'onlineSavingsMsg'
Learn More

Gibco™ Human Lin28-TAT Recombinant Protein, PeproTech®

Product Code. p-7246035
Click to view available options
Quantity:
1 mg
100 μg
25 μg
250 μg
5 μg
500 μg
Unit Size:
100µg
1mg
250µg
25µg
500µg
5µg
This item is not returnable. View return policy

Product Code. 17821763

Brand: Gibco™ 11006100UG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Recombinant Protein

Recombinant Human Lin28-TAT is a 24.4 kDa protein containing 222 amino acid residues, including a 13-residue C-terminal TAT peptide. This product is shipped at ambient temperature. For storage, handling and reconstitution information, please see the lot-specific Certificate of Analysis

Lin28 is a RNA-binding protein that belongs to a diverse family of structurally-related transcription factors. Lin28 is found abundantly in embryonic stem cells (ESCs), and to a lesser extent in placenta and testis. Lin28 has been shown to block let-7 microRNA processing and maturation, a necessary step in the differentiation of stem cells and certain cancer cell lines. Together with Sox2, Oct4, and Nanog, Lin28 can induce the reprogramming of primary human fibroblasts to a pluripotent state. Lin28 and other regulatory proteins can be introduced into cells by DNA transfection, viral infection, or microinjection. Protein transduction using TAT fusion proteins represents an alternative methodology for introducing proteins into primary, as well as transformed, cells.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number Q9H9Z2
For Use With (Application) Functional Assay
Formulation protein with no preservative
Gene ID (Entrez) 79727
Molecular Weight (g/mol) 24.4 kDa
Name Human Lin28-TAT
Quantity 100 μg
Source E. coli
Regulatory Status RUO
Endotoxin Concentration <1 EU/ μg
Gene Alias LIN28A
Common Name LIN28A-TAT
Gene Symbol LIN28A
Biological Activity Measured by its ability to induce fluorescence in Lin28 reporter cells (293 cells transfected with fluorescent protein genes under Lin28 control). Optimum activity was achieved (at 20 ug/ml after incubation for 72 hr.)
Conjugate Unconjugated
Recombinant Recombinant
Protein Tag TAT-fusion
Sequence GPSVSNQQFA GGCAKAAEEA PEEAPEDAAR AADEPQLLHG AGICKWFNVR MGFGFLSMTA RAGVALDPPV DVFVHQSKLH MEGFRSLKEG EAVEFTFKKS AKGLESIRVT GPGGVFCIGS ERRPKGKSMQ KRRSKGDRCY NCGGLDHHAK ECKLPPQPKK CHFCQSISHM VASCPLKAQQ GPSAQGKPTY FREEEEEIHS PTLLPEAQNG GYGRKKRRQR RR
Content And Storage -20°C
Expression System E. coli
Form Lyophilized
Purity or Quality Grade ≥ 90% by SDS-PAGE gel and HPLC analyses.
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.