missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human LIN-28B Antibody, R&D Systems™

Sheep Polyclonal Antibody
Brand: R&D Systems AF7726-SP
This item is not returnable.
View return policy
Description
LIN-28B Polyclonal specifically detects LIN-28B in Human samples. It is validated for Western Blot, Immunohistochemistry.
Specifications
| LIN-28B | |
| Polyclonal | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| CSDD2, CSDD2protein lin-28 homolog B, FLJ16517, lin-28 homolog B (C. elegans), Lin-28.2, lin-28B, LIN28B | |
| Sheep | |
| Affinity Purified | |
| RUO | |
| 389421 | |
| Sterile PBS to a final concentration of 0.2 mg/mL. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
| Western Blot, Immunohistochemistry | |
| LYOPH | |
| Western Blot 0.5 ug/mL, Immunohistochemistry 5-15 ug/mL | |
| Q6ZN17 | |
| LIN28B | |
| E. coli-derived recombinant human LIN-28B Val83-Thr250 Accession # Q6ZN17 | |
| 25 μg | |
| Primary | |
| Detects human LIN-28B in direct ELISAs and Western blots. In direct ELISAs, less than 4% cross-reactivity with recombinant human LIN-28A is observed. | |
| Human | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction