missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human LIN-28A Antibody, R&D Systems™

Goat Polyclonal Antibody has been used in 12 publications
Brand: R&D Systems AF3757-SP
This item is not returnable.
View return policy
Description
LIN-28A Polyclonal specifically detects LIN-28A in Human samples. It is validated for Western Blot, Immunocytochemistry.
Specifications
| LIN-28A | |
| Polyclonal | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| CSDD1, CSDD1lin-28A, FLJ12457, LIN28, LIN-28, lin-28 homolog (C. elegans), lin-28 homolog A (C. elegans), Lin-28A, LIN28A, LIN28RNA-binding protein LIN-28, Tex17, ZCCHC1, ZCCHC1protein lin-28 homolog A, Zinc finger CCHC domain-containing protein 1, zinc finger, CCHC domain containing 1 | |
| Goat | |
| Affinity Purified | |
| RUO | |
| 79727 | |
| Reconstitute at 0.2 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
| Western Blot, Immunocytochemistry | |
| LYOPH | |
| Western Blot 0.1 ug/mL, Immunocytochemistry 5-15 ug/mL | |
| Q9H9Z2 | |
| LIN28A | |
| E. coli-derived recombinant human LIN-28A Met1-Asn209 Accession # Q9H9Z2 | |
| 25 μg | |
| Primary | |
| Detects human LIN-28A in direct ELISAs and Western blots. | |
| Human | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction