missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human LILRB4/CD85k/ILT3 Fluorescein-conjugated Antibody, R&D Systems™

Mouse Monoclonal Antibody has been used in 3 publications
£155.00 - £300.00
Specifications
| Antigen | ILT3/CD85k/LILRB4 |
|---|---|
| Clone | 293623 |
| Dilution | Flow Cytometry 10 uL/10^6 cells |
| Applications | Flow Cytometry |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
18777368
![]() |
R&D Systems
FAB24251F-025 |
25 Tests |
£155.00
25 tests |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
18729806
![]() |
R&D Systems
FAB24251F-100 |
100 Tests |
£300.00
100 tests |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
ILT3/CD85k/LILRB4 Monoclonal specifically detects ILT3/CD85k/LILRB4 in Human samples. It is validated for Flow Cytometry.Specifications
| ILT3/CD85k/LILRB4 | |
| Flow Cytometry 10 uL/10^6 cells | |
| Monoclonal | |
| Purified | |
| RUO | |
| Supplied in a saline solution containing BSA and Sodium Azide. with Sodium Azide | |
| CD85 antigen-like family member K, CD85k, CD85k antigen, HM18, ILT3, ILT-3, ILT3CD85K, leukocyte immunoglobulin-like receptor, subfamily B (with TM and ITIM domains), LIR5, LIR5LILRB5, LIR-5subfamily B, member 4, member 4 | |
| LILRB4 | |
| IgG2a | |
| Protein A or G purified from hybridoma culture supernatant | |
| Detects human ILT3/CD85k in direct ELISAs and Western blots. In direct ELISAs, approximately 50% cross-reactivity with with recombinant human (rh) ILT5 is observed. No cross-reactivity with rhILT1, rhILT2, rhILT4, rhILT7, rhILT11, rhLIR-6 or rhLIR-8 is observed. |
| 293623 | |
| Flow Cytometry | |
| Fluorescein | |
| Mouse | |
| Human | |
| ABM86208 | |
| 11006 | |
| Mouse myeloma cell line NS0-derived recombinant human ILT3/CD85k Pro17-His257 Accession # ABM86208 | |
| Primary | |
| Protect from light. Do not freeze. 12 months from date of receipt, 2 to 8 degreesC as supplied. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title