missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human LILRB4/CD85k/ILT3 Antibody, R&D Systems™

Mouse Monoclonal Antibody
£168.00 - £393.00
Specifications
| Antigen | ILT3/CD85k/LILRB4 |
|---|---|
| Clone | 293622 |
| Concentration | LYOPH |
| Dilution | Western Blot 1 ug/mL |
| Applications | Western Blot |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
18754438
![]() |
R&D Systems
MAB2425-SP |
25 μg |
£168.00
25µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18109450
|
R&D Systems
MAB2425 |
100 μg |
£393.00
100µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
ILT3/CD85k/LILRB4 Monoclonal specifically detects ILT3/CD85k/LILRB4 in Human samples. It is validated for Western Blot.Specifications
| ILT3/CD85k/LILRB4 | |
| LYOPH | |
| Western Blot | |
| Unconjugated | |
| Mouse | |
| Human | |
| ABM86208 | |
| 11006 | |
| Mouse myeloma cell line NS0-derived recombinant human LILRB4/CD85k/ILT3 Pro17-His257 Accession # ABM86208 | |
| Primary | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Detects human LILRB4/CD85k/ILT3 in direct ELISAs and Western blots. In Western blots, no cross-reactivity with recombinant human (rh) ILT2, 4 5, 6, rhLIR6, or rhLIR8 is observed. |
| 293622 | |
| Western Blot 1 ug/mL | |
| Monoclonal | |
| Purified | |
| RUO | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| CD85 antigen-like family member K, CD85k, CD85k antigen, HM18, ILT3, ILT-3, ILT3CD85K, leukocyte immunoglobulin-like receptor, subfamily B (with TM and ITIM domains), LIR5, LIR5LILRB5, LIR-5subfamily B, member 4, member 4 | |
| LILRB4 | |
| IgG2a | |
| Protein A or G purified from hybridoma culture supernatant | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title