missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human LILRB3/CD85a/ILT5 APC-conjugated Antibody, R&D Systems™

Mouse Monoclonal Antibody
£180.00 - £456.00
Specifications
| Antigen | ILT5/CD85a/LILRB3 |
|---|---|
| Clone | 222821 |
| Dilution | Flow Cytometry 10 uL/10^6 cells |
| Applications | Flow Cytometry |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
18711238
![]() |
R&D Systems
FAB1806A-025 |
25 Tests |
£180.00
25 tests |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
18788016
![]() |
R&D Systems
FAB1806A-100 |
100 Tests |
£456.00
100 tests |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
ILT5/CD85a/LILRB3 Monoclonal specifically detects ILT5/CD85a/LILRB3 in Human samples. It is validated for Flow Cytometry.Specifications
| ILT5/CD85a/LILRB3 | |
| Flow Cytometry 10 uL/10^6 cells | |
| Monoclonal | |
| Purified | |
| RUO | |
| Supplied in a saline solution containing BSA and Sodium Azide. with Sodium Azide | |
| CD85 antigen-like family member A, CD85a, CD85a antigen, HL9Immunoglobulin-like transcript 5, ILT5, ILT-5, ILT5CD85a, Leukocyte immunoglobulin-like receptor 3, leukocyte immunoglobulin-like receptor subfamily B member 3, leukocyte immunoglobulin-like receptor, subfamily B (with TM and ITIM domains), LIR3, LIR3CD85A, LIR-3MGC138403, member 3, Monocyte inhibitory receptor HL9, PIRB | |
| LILRB3 | |
| IgG2a | |
| Protein A or G purified from hybridoma culture supernatant | |
| Detects human ILT5/CD85a in direct ELISAs and Western blots. In direct ELISAs, no cross-reactivity with recombinant human (rh) ILT1, rhILT2, rhILT3, rhILT4, rhILT6, rhILT7 or rhILT11 is observed. |
| 222821 | |
| Flow Cytometry | |
| APC | |
| Mouse | |
| Human | |
| AAB68668 | |
| 11025 | |
| Mouse myeloma cell line NS0-derived recombinant human ILT5/CD85a Gly24-Glu443 Accession # AAB68668 | |
| Primary | |
| Protect from light. Do not freeze. 12 months from date of receipt, 2 to 8 degreesC as supplied. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title