missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human LILRB3/CD85a/ILT5 Antibody, R&D Systems™
Mouse Monoclonal Antibody has been used in 6 publications
Brand: R&D Systems MAB1806-100
This item is not returnable.
View return policy
Description
ILT5/CD85a/LILRB3 Monoclonal specifically detects ILT5/CD85a/LILRB3 in Human samples. It is validated for Western Blot, Flow Cytometry, CyTOF-ready.
Specifications
| ILT5/CD85a/LILRB3 | |
| Monoclonal | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| CD85 antigen-like family member A, CD85a, CD85a antigen, HL9Immunoglobulin-like transcript 5, ILT5, ILT-5, ILT5CD85a, Leukocyte immunoglobulin-like receptor 3, leukocyte immunoglobulin-like receptor subfamily B member 3, leukocyte immunoglobulin-like receptor, subfamily B (with TM and ITIM domains), LIR3, LIR3CD85A, LIR-3MGC138403, member 3, Monocyte inhibitory receptor HL9, PIRB | |
| Mouse | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| 11025 | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
| IgG2a |
| Western Blot, Flow Cytometry, CyTOF | |
| 222821 | |
| Western Blot 1 ug/mL, Flow Cytometry 0.25 ug/10^6 cells, CyTOF-ready | |
| AAB68668 | |
| LILRB3 | |
| Mouse myeloma cell line NS0-derived recombinant human LILRB3/CD85a/ILT5 Gly24-Glu443 Accession # AAB68668 | |
| 100 μg | |
| Primary | |
| Detects human LILRB3/CD85a/ILT5 in direct ELISAs and Western blots. In direct ELISAs, no cross-reactivity with recombinant human (rh) ILT1, rhILT2, rhILT3, rhILT4, rhILT6, rhILT7 or rhILT11 is observed. | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction