missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human LILRA1/LILRB1 Antibody, R&D Systems™

Mouse Monoclonal Antibody
Brand: R&D Systems MAB30851-SP
Dieser Artikel kann nicht zurückgegeben werden.
Rückgaberichtlinie anzeigen
Beschreibung
LILRA1/LILRB1 Monoclonal specifically detects LILRA1/LILRB1 in Human samples. It is validated for Flow Cytometry, CyTOF-ready.
Spezifikation
| LILRA1/LILRB1 | |
| Monoclonal | |
| LYOPH | |
| Flow Cytometry 0.25 ug/10^6 cells, CyTOF-ready | |
| O75019 | |
| LILRA1 | |
| Mouse myeloma cell line NS0-derived recombinant human LILRA1 Pro17-Asn461 Accession # O75019 | |
| 25 μg | |
| Primary | |
| Detects human LILRA1 in direct ELISAs. In direct ELISAs, 100% cross-reactivity with recombinant human (rh) LILRB1 is observed and no cross-reactivity rhILT3, 4, 5, 6, rhLIR-7 or -8 is observed. | |
| Human | |
| Purified |
| Flow Cytometry, CyTOF | |
| 586326 | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| CD85I, CD85j, ILT2, LIR1, LIR6, LIR-6, MIR7 | |
| Mouse | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| 11024 | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
| IgG2b |
Berichtigung von Produktinhalten
Bitte geben Sie uns Ihr Feedback zu den Produktinhalten, indem Sie das folgende Formular ausfüllen.
Name des Produkts
For Research Use Only
Haben Sie Verbesserungsvorschläge?Übermitteln Sie eine inhaltliche Korrektur