missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems™ Human LIGHT/TNFSF14 Quantikine ELISA Kit

Product Code. 18702344
Change view
Click to view available options
Quantity:
1 Kit
Unit Size:
1 set
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18702344 1 Kit 1 set
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18702344 Supplier R&D Systems™ Supplier No. DLIT00

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Human LIGHT/TNFSF14 Quantikine ELISA Kit from the most referenced ELISA manufacturer. Highly validated for accurate quantitation and long-term reproducibility.

The Human LIGHT/TNFSF14 Quantikine ELISA Kit is a Solid Phase Sandwich ELISA that quantifies human LIGHT/TNFSF14 in cell culture supernates (50 ul), serum (50 ul), edta plasma (50 ul), heparin plasma (50 ul), saliva (25 ul).
TRUSTED_SUSTAINABILITY

Specifications

Assay Range 31.20 to 2000 pg/mL
Assay Sensitivity 16.5 pg/mL
Conjugate HRP
Product Type Solid Phase Sandwich ELISA
Sample Type Cell Culture Supernatant, Plasma (EDTA, Heparin), Saliva, Serum
Specificity Natural and Recombinant Human LIGHT
For Use With (Application) ELISA
Gene ID (Entrez) 8740
Gene Symbol TNFSF14
Quantity 1 Kit
Sample Volume Cell Culture Supernates (50uL),Serum (50uL),EDTA Plasma (50uL),Heparin Plasma (50uL),Saliva (25uL)
Storage Requirements Store the unopened product at 2°C to 8°C. Do not use past expiration date.
Synonym CD258, CD258 antigen, delta transmembrane LIGHT, Herpes virus entry mediator ligand, Herpesvirus entry mediator ligand, HVEM-L, HVEMLherpesvirus entry mediator-ligand, ligand for herpesvirus entry mediator, LIGHTherpesvirus entry mediator A, LTg, TNFSF14, TR2, tumor necrosis factor (ligand) superfamily, member 14, tumor necrosis factor ligand superfamily member 14, tumor necrosis factor receptor-like 2, tumor necrosis factor superfamily member LIGHT
Target LIGHT/TNFSF14
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.