missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human LIFR alpha Antibody, R&D Systems™

Mouse Monoclonal Antibody has been used in 2 publications
Brand: R&D Systems MAB249
This item is not returnable.
View return policy
Description
LIFR alpha Monoclonal specifically detects LIFR alpha in Human samples. It is validated for Western Blot, Flow Cytometry, CyTOF-ready.
Specifications
| LIFR alpha | |
| Monoclonal | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| CD118, CD118 antigen, FLJ98106, FLJ99923, leukemia inhibitory factor receptor, leukemia inhibitory factor receptor alpha, LIF R alpha, LIF R beta, LIF receptor, LIFR, LIF-R, LIFRa, SJS2, STWS, SWS | |
| Mouse | |
| Protein A or G purified from ascites | |
| RUO | |
| 3977 | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
| IgG1 |
| Western Blot, Flow Cytometry, CyTOF | |
| 32953 | |
| Western Blot 1 ug/mL, Flow Cytometry 2.5 ug/10^6 cells, CyTOF-ready | |
| P42702 | |
| LIFR | |
| S. frugiperda insect ovarian cell line Sf 21-derived recombinant human LIF R alpha Gln45-Ser833 Accession # P42702 | |
| 500 μg | |
| Primary | |
| Detects human LIF R alpha in direct ELISAs and Western blots. | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction