missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human LIF Antibody, R&D Systems™

Mouse Monoclonal Antibody
Brand: R&D Systems MAB2502-SP
This item is not returnable.
View return policy
Description
LIF Monoclonal antibody specifically detects LIF in Human samples. It is validated for ELISA
Specifications
| LIF | |
| Monoclonal | |
| Unconjugated | |
| P15018 | |
| Mouse | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| 3976 | |
| Human | |
| Purified |
| ELISA | |
| 945842 | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. Small pack size (SP) is supplied either lyophilized or as a 0.2 μm filtered solution in PBS. | |
| CDF, D factor, DIA, differentiation inhibitory activity, differentiation stimulating factor, Differentiation-stimulating factor, Emfilermin, HILDAcholinergic differentiation factor, leukemia inhibitory factor, leukemia inhibitory factor (cholinergic differentiation factor), Melanoma-derived LPL inhibitor, MLPLI | |
| Human embryonic kidney cell, HEK293-derived human LIF, Met1-Phe202, Accession # P15018 | |
| 25 μg | |
| Primary | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20°C to -70°C as supplied. 1 month, 2°C to 8°C under sterile conditions after reconstitution. 6 months, -20°C to -70°C under sterile conditions after reconstitution. | |
| IgG2b |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction