missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human Lgr6 Antibody, R&D Systems™

Mouse Monoclonal Antibody
Brand: R&D Systems MAB84581-SP
This item is not returnable.
View return policy
Description
Lgr6 Monoclonal specifically detects Lgr6 in Human samples. It is validated for Flow Cytometry, Blockade of Receptor-ligand Interaction, Immunocytochemistry, CyTOF-ready.
Specifications
| Lgr6 | |
| Monoclonal | |
| Unconjugated | |
| Lyophilized from a 0.2μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2μm filtered solution in PBS. | |
| FLJ14471, gonadotropin receptor, GPCR, leucine-rich repeat containing G protein-coupled receptor 6, leucine-rich repeat-containing G protein-coupled receptor 6, leucine-rich repeat-containing G-protein coupled receptor 6, VTS20631 | |
| NS0 mouse myeloma cell line transfected with human Lgr6 Met1-Val967 Accession # Q9HBX8 | |
| 25 μg | |
| 59352 | |
| Reconstitute at 0.5mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
| IgG2a |
| Flow Cytometry, Blocking Assay, Immunocytochemistry, CyTOF | |
| 918726 | |
| Flow Cytometry 0.25 ug/10^6 cells, Blockade of Receptor-ligand Interaction 1 μg/mL, Immunocytochemistry 8-25 μg/mL, CyTOF-ready | |
| Q9HBX8 | |
| Mouse | |
| Protein A or G purified from hybridoma culture supernatant | |
| Primary | |
| Detects human Lgr6 in direct ELISAs. | |
| Human | |
| Supernatant |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction