missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human Lgr5/GPR49 Antibody, R&D Systems™

Mouse Monoclonal Antibody has been used in 2 publications
Brand: R&D Systems MAB8078
This item is not returnable.
View return policy
Description
Lgr5/GPR49 Monoclonal specifically detects Lgr5/GPR49 in Human samples. It is validated for Flow Cytometry, Agonist Activity, Immunocytochemistry, CyTOF-ready.
Specifications
| Lgr5/GPR49 | |
| Monoclonal | |
| Unconjugated | |
| O75473 | |
| LGR5 | |
| Chinese hamster ovary cell line CHO-derived recombinant human Lgr5/GPR49 Met1-Ile560 Accession # O75473 | |
| 100 μg | |
| 8549 | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
| IgG2a |
| Flow Cytometry, Agonist Activity, Immunocytochemistry, CyTOF | |
| 707042 | |
| Flow Cytometry 2.5 ug/10^6 cells, Agonist Activity, Immunocytochemistry 8-25 ug/mL, CyTOF-ready | |
| FEX, G protein-coupled receptor 49, GPR49, GPR49G-protein coupled receptor HG38, GPR67, G-protein coupled receptor 49, G-protein coupled receptor 67, GRP49, HG38, leucine-rich repeat containing G protein-coupled receptor 5, leucine-rich repeat-containing G protein-coupled receptor 5, leucine-rich repeat-containing G-protein coupled receptor 5, MGC117008, orphan G protein-coupled receptor HG38 | |
| Mouse | |
| Protein A or G purified from hybridoma culture supernatant | |
| Primary | |
| Detects human Lgr5/GPR49 in ELISA. Stains human and mouse Lgr5 transfected cells but not irrelevant transfectants in Flow Cytometry and Immunocytochemistry. This antibody, also known as “RD42”, has been found to detect an epitope in C-terminal LRR cap of LGR5 (Ref. 1). | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction