missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human Lgr4/GPR48 APC-conjugated Antibody, R&D Systems™

Mouse Monoclonal Antibody
Brand: R&D Systems FAB7750A-100
This item is not returnable.
View return policy
Description
Lgr4/GPR48 Monoclonal specifically detects Lgr4/GPR48 in Human samples. It is validated for Flow Cytometry.
Specifications
| Lgr4/GPR48 | |
| Monoclonal | |
| APC | |
| Supplied in a saline solution containing BSA and Sodium Azide. with Sodium Azide | |
| GPR48, GPR48G protein-coupled receptor 48, G-protein coupled receptor 48, leucine-rich repeat containing G protein-coupled receptor 4 | |
| Mouse | |
| Protein A or G purified from hybridoma culture supernatant | |
| Primary | |
| Detects human Lgr4/GPR48 in Lgr4-transfected cell-based ELISA. Stains human Lgr4/GPR48 transfected but not irrelevant transfected cells in flow cytometry. | |
| Protect from light. Do not freeze. 12 months from date of receipt, 2 to 8°C as supplied. | |
| IgG2b |
| Flow Cytometry | |
| 852229 | |
| Flow Cytometry 10 μL/106 cells | |
| Q9BXB1 | |
| LGR4 | |
| NS0 mouse myeloma cell line transfected with human Lgr4/GPR48, Met1-Asp951, Accession No. Q9BXB1 | |
| 100 Tests | |
| 55366.0 | |
| Human | |
| Supernatant |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction