missing translation for 'onlineSavingsMsg'
Learn More

Human Lgr4/GPR48 Antibody, R&D Systems™
GREENER_CHOICE

Product Code. 18740718
Change view
Click to view available options
Quantity:
25 μg
Unit Size:
25µg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18740718 25 μg 25µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18740718 Supplier R&D Systems Supplier No. MAB7750SP

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

Lgr4/GPR48 Monoclonal specifically detects Lgr4/GPR48 in Human samples. It is validated for Flow Cytometry, Blockade of Receptor-ligand Interaction, Immunocytochemistry, CyTOF-ready.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Lgr4/GPR48
Applications Flow Cytometry, Blockade of Receptor-ligand Interaction, Immunocytochemistry, CyTOF
Classification Monoclonal
Clone 852229
Conjugate Unconjugated
Dilution Flow Cytometry 0.25 ug/10^6 cells, Blockade of Receptor-ligand Interaction 2.5 ug/mL, Immunocytochemistry 8-25 ug/mL, CyTOF-ready
Formulation Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
Gene Accession No. Q9BXB1
Gene Alias GPR48, GPR48G protein-coupled receptor 48, G-protein coupled receptor 48, leucine-rich repeat containing G protein-coupled receptor 4
Gene Symbols LGR4
Host Species Mouse
Immunogen NS0 mouse myeloma cell line transfected with human Lgr4/GPR48 Met1-Asp951 Accession # Q9BXB1
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 25 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 55366
Test Specificity Detects human Lgr4/GPR48 in Lgr4-transfected cell-based ELISA. Stains human Lgr4/GPR48 transfected but not irrelevant transfected cells in flow cytometry.
Reconstitution Reconstitute at 0.5 mg/mL in sterile PBS.
Target Species Human
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Form Purified
Isotype IgG2b
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.