missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Human LGI2 (aa 134-259) Control Fragment Recombinant Protein

Product Code. 30200737
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
This item is not returnable. View return policy

Product Code. 30200737

Brand: Invitrogen™ RP90365

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Recombinant Protein

Highest antigen sequence indentity to the following orthologs: Mouse (99%), Rat (99%). This recombinant protein control fragment may be used for blocking experiments with the corresponding antibody, PA5-53555 (PA5-53555. In IHC/ICC and WB experiments, we recommend a 100x molar excess of the protein fragment control based on the concentration and the molecular weight. Pre-incubate the antibody-protein control fragment mixture for 30 min at room temperature.

The leucine-rich, glioma inactivated gene 2 (LGI2) is a member of the LGI family in which LGI1 is the exemplar. The LGI family consists of four of highly related proteins containing leucine-rich repeats (LRRs) which are highly similar to other transmembrane signaling molecules and receptors. LGI1 has been identified as a candidate tumor suppressor gene for glioma and plays a role in autodominant lateral temporal epilepsy (ADTLE), an epileptic syndrome characterized by focal seizures with predominant auditory symptoms. Despite its high homology with LGI1 and similar pattern of expression, mutations in LGI2 have not been found to be associated with ADTLE.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number Q8N0V4
Concentration ≥5.0 mg/mL
For Use With (Application) Blocking Assay, Control
Formulation 1 M urea, PBS with no preservative; pH 7.4
Gene ID (Entrez) 55203
Name Human LGI2 (aa 134-259) Control Fragment
Quantity 100 μL
Regulatory Status RUO
Gene Alias Kiaa1916; leucine rich repeat LGI family member 2; leucine-rich glioma-inactivated protein 2; Leucine-rich repeat LGI family member 2; leucine-rich repeat LGI family, member 2; leucine-rich, glioma inactivated 2; LGI1-like protein 2; Lgi2; LGIL2; mKIAA1916
Common Name LGI2
Gene Symbol Lgi2
Conjugate Unconjugated
Species Human
Recombinant Recombinant
Protein Tag His-ABP-tag
Sequence DLTHLSLANNHIKALPRDVFSDLDSLIELDLRGNKFECDCKAKWLYLWLKMTNSTVSDVLCIGPPEYQEKKLNDVTSFDYECTTTDFVVHQTLPYQSVSVDTFNSKNDVYVAIAQPSMENCMVLEW
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Expression System E. coli
Form Liquid
Purity or Quality Grade >80% by SDS-PAGE and Coomassie blue staining
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.