missing translation for 'onlineSavingsMsg'
Learn More

Proteintech™ Human LGALS3BP ELISA Kit

Product Code. 16821566 Shop All Proteintech Products
Click to view available options
Target Species:
Human
Unit Size:
96 tests
This item is not returnable. View return policy

Product Code. 16821566

Brand: Proteintech™ KE0015596T

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Solid phase sandwich ELISA used to detect and quantify protein levels of endogenous LGALS3BP

  • Recognizes human LGALS3BP
  • Galactins are a family of beta-galactoside-binding proteins implicated in modulating cell-cell and cell-matrix interactions
    • Galectin-3-binding protein (LGALS3BP, 90K, Mac-2 BP) is a secreted glycoprotein binds galectin-3, galectin-1, beta1 integrins, collagens, fibronectin
    • Implicated in tumor metastatic processes and other cell adhesion and immune functions
    • Found elevated in serum of patients with cancer and in those infected by HIV
    • Found in breast milk, semen, saliva, urine, tears and serum

Process:

  • Antibody specific for LGALS3BP pre-coated onto microwells
  • LGALS3BP protein in samples captured by the coated antibody after incubation
  • After extensive washing, another antibody of biotinylated specific for human LGALS3BP is added to detect captured human LGALS3BP protein
  • For signal development, Streptavidin-HRP is added, followed by Tetramethyl-benzidine (TMB) reagent
  • Solution containing sulfuric acid used to stop color development and color intensity
    • Proportional to quantity of bound protein, measurable at 450 nm with correction wavelength set at 630 nm

Specifications

Accession Number BC002403
Assay Range 15.6 to 1000 pg/mL
Assay Sensitivity 5.2 pg/mL
Certifications/Compliance ISO 13485
Detection Method Sandwich ELISA, Double Antibody
Immunoassay Kit Format Coated ELISA Kit
Sample Type Plasma, Serum
Substrate DAB
Target Species Human
Gene ID (Entrez) 3959
Host Species Human
Immunoassay Kit Grade Solid
Interassay CV <10%
Intraassay CV <10%
Shipping Condition Blue ice
Storage Requirements -20°C to +4°C
Total Assay Time 4.5 hr.
UniProt ID Q08380
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • 96TEST Human ANGPTL4 ELISA Kit
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 1
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H302-Harmful if swallowed.
  • H315-Causes skin irritation.
  • H318-Causes serious eye damage.
  • H319-Causes serious eye irritation.
  • H332-Harmful if inhaled.
  • H335-May cause respiratory irritation.
Precautionary Statement
  • P201-Obtain special instructions before use.
  • P202-Do not handle until all safety precautions have been read and understood.
  • P234-Keep only in original container.
  • P264-Wash skin thoroughly after handling.
  • P270-Do not eat, drink or smoke when using this product.
  • P273-Avoid release to the environment.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P308+P313-IF exposed or concerned: Get medical advice/attention.
  • P330-Rinse mouth.
  • P362-Take off contaminated clothing.
  • P405-Store locked up.
  • P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Supplemental information
  • MIXTURE LIST-Contains : Sulphuric Acid, Tetramethylbenzidine
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.