missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human Leucyl-cystinyl Aminopeptidase/LNPEP Antibody, R&D Systems™

Sheep Polyclonal Antibody
Brand: R&D Systems AF6386-SP
This item is not returnable.
View return policy
Description
Leucyl-cystinyl Aminopeptidase/LNPEP Polyclonal specifically detects Leucyl-cystinyl Aminopeptidase/LNPEP in Human samples. It is validated for Western Blot, Immunoprecipitation.
Specifications
| Leucyl-cystinyl Aminopeptidase/LNPEP | |
| Polyclonal | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| angiotensin IV receptor, AT (4) receptor, CAP, Cystinyl aminopeptidase, EC 3.4.11, EC 3.4.11.3, Insulin-regulated membrane aminopeptidase, Insulin-responsive aminopeptidase, IRAP, leucyl/cystinyl aminopeptidase, leucyl-cystinyl aminopeptidase, Leucylcystinyl Aminopeptidase, LNPEP, Otase, Oxytocinase, Placental leucine aminopeptidase, PLAP, P-LAP, P-LAPinsulin-regulated aminopeptidase, vasopressinase | |
| Sheep | |
| Affinity Purified | |
| RUO | |
| 4012 | |
| Sterile PBS to a final concentration of 0.2 mg/mL. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
| Western Blot, Immunoprecipitation | |
| LYOPH | |
| Western Blot 1 ug/mL, Immunoprecipitation 25 ug/mL | |
| Q9UIQ6 | |
| LNPEP | |
| Chinese hamster ovary cell line CHO-derived recombinant human Leucyl-cystinyl Aminopeptidase/LNPEP Arg133-Gly552 Accession # Q9UIQ6 | |
| 25 μg | |
| Primary | |
| Detects human Leucyl-cystinyl Aminopeptidase/LNPEP in direct ELISAs and Western blots. In direct ELISAs, less than 2% cross-reactivity with recombinant human (rh) ENPEP, rhLRAP, and rhARTS1 is observed. | |
| Human | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction