missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Human LETM2 (aa 356-421) Control Fragment Recombinant Protein

Product Code. 30195049
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
This item is not returnable. View return policy

Product Code. 30195049

Brand: Invitrogen™ RP93882

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Recombinant Protein

Highest antigen sequence indentity to the following orthologs: Mouse (71%), Rat (71%). This recombinant protein control fragment may be used for blocking experiments with the corresponding antibody, PA5-55209 (PA5-55209. In IHC/ICC and WB experiments, we recommend a 100x molar excess of the protein fragment control based on the concentration and the molecular weight. Pre-incubate the antibody-protein control fragment mixture for 30 min at room temperature.

LETM2 contains 1 LETM1 domain. The function of the LETM1 protein remains unknown.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number Q2VYF4
Concentration ≥5.0 mg/mL
For Use With (Application) Blocking Assay, Control
Formulation 1 M urea, PBS with no preservative; pH 7.4
Gene ID (Entrez) 137994
Name Human LETM2 (aa 356-421) Control Fragment
Quantity 100 μL
Regulatory Status RUO
Gene Alias 6030453H13; D030041N04Rik; LETM1 and EF-hand domain-containing protein 2; LETM1 domain-containing protein LETM2, mitochondrial; LETM2; LETM2S; leucine zipper and EF-hand containing transmembrane protein 2; leucine zipper-, EF-hand motif- containing transmembrane protein 2; leucine zipper-, EF-hand motif- containing transmembrane protein 2 S; leucine zipper-EF-hand containing transmembrane protein 1-like protein; leucine zipper-EF-hand containing transmembrane protein 2; Leucine zipper-EF-hand-containing transmembrane protein 1-like
Common Name LETM2
Gene Symbol LETM2
Conjugate Unconjugated
Species Human
Recombinant Recombinant
Protein Tag His-ABP-tag
Sequence GLTEEQLRQQLTEWQDLHLKENVPPSLLLLSRTFYLIDVKPKPIEIPLSGEAPKTDILVELPTFTE
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Expression System E. coli
Form Liquid
Purity or Quality Grade >80% by SDS-PAGE and Coomassie blue staining
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.