missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human Leptin R Antibody, R&D Systems™

Mouse Monoclonal Antibody has been used in 4 publications
Brand: R&D Systems MAB867-100
This item is not returnable.
View return policy
Description
Leptin R Monoclonal specifically detects Leptin R in Human samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, CyTOF-ready.
Specifications
| Leptin R | |
| Monoclonal | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| B219, CD295, CD295 antigen, DB, DKFZp686B1731, huB219, LEPR, LEP-R, leptin receptor, LeptinR, OB R, OB receptor, OB-R, OBRCD295 | |
| Mouse | |
| Protein A or G purified from ascites | |
| RUO | |
| 3953 | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
| IgG2b |
| Western Blot, Flow Cytometry, Immunohistochemistry, CyTOF | |
| 52263 | |
| Western Blot 1 ug/mL, Flow Cytometry 2.5 ug/10^6 cells, Immunohistochemistry 8-25 ug/mL, CyTOF-ready | |
| P48357 | |
| LEPR | |
| Mouse myeloma cell line NS0-derived recombinant human Leptin R Phe22-Asp839 (predicted) Accession # P48357 | |
| 100 μg | |
| Primary | |
| Detects human Leptin R in direct ELISAs and Western blots. In Western blots, cross reactivity with recombinant mouse Leptin R is observed. | |
| Human | |
| Ascites |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction