missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human Leptin R Antibody, Novus Biologicals™

Mouse Monoclonal Antibody
Brand: R&D Systems MAB86713-SP
This item is not returnable.
View return policy
Description
Leptin R Monoclonal antibody specifically detects Leptin R in nan samples. It is validated for ELISA
Specifications
| Leptin R | |
| Monoclonal | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied either lyophilized or as a 0.2 μm filtered solution in PBS. | |
| B219, CD295, CD295 antigen, DB, DKFZp686B1731, huB219, LEPR, LEP-R, leptin receptor, LeptinR, OB R, OB receptor, OB-R, OBRCD295 | |
| Mouse myeloma cell line NS0-derived recombinant human Leptin R, Accession # P48357 | |
| 25 μg | |
| Primary | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Purified |
| ELISA | |
| 52276 | |
| ELISA | |
| P48357 | |
| Mouse | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| 3953 | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 °C as supplied. 1 month, 2 to 8 °C under sterile conditions after reconstitution. 6 months, -20 to -70 °C under sterile conditions after reconstitution. | |
| IgG2a |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction